MAT333116457 Matrix Scientific

2-Benzoylcyclohexanecarboxylic acid

  • MDL number MFCD07775665
  • Cas Number : 6939-99-7
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061277.html

Safety Information

MSDS http://www.matrixscientific.com/061277.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart