MAT333116616 Matrix Scientific

6-(2,3-Dihydro-1H-inden-5-yloxy)nicotinic acid

  • MDL number MFCD12197752
  • Cas Number : 1042778-06-2
  • Molecular Weight : 255.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/061436.html

Safety Information

MSDS http://www.matrixscientific.com/061436.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart