MAT333116621 Matrix Scientific

Pyridine-2,5-dicarboxamide

  • MDL number MFCD00457415
  • Cas Number : 4663-96-1
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061441.html

Safety Information

MSDS http://www.matrixscientific.com/061441.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart