MAT333116667 Matrix Scientific

4-Chloro-1-methyl-1H-pyrazole-5-carbonyl chloride

  • MDL number MFCD03421434
  • Cas Number : 157142-50-2
  • Molecular Weight : 179.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061487.html

Safety Information

MSDS http://www.matrixscientific.com/061487.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart