MAT333116892 Matrix Scientific

8-Chloro-2-(3-methylphenyl)quinoline-4-carbonyl chloride

  • MDL number MFCD03422815
  • Cas Number : 1160263-53-5
  • Molecular Weight : 316.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061712.html

Safety Information

MSDS http://www.matrixscientific.com/061712.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart