MAT333117097 Matrix Scientific

2-Methyl-2-(2-methylphenoxy)propanoyl chloride

  • MDL number MFCD11939559
  • Cas Number : 116762-25-5
  • Molecular Weight : 212.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061917.html

Safety Information

MSDS http://www.matrixscientific.com/061917.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart