MAT333117112 Matrix Scientific

2-(2,4-Dichlorophenoxy)-2-methylpropanoyl chloride

  • MDL number MFCD12197862
  • Cas Number : 41711-48-2
  • Molecular Weight : 267.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061932.html

Safety Information

MSDS http://www.matrixscientific.com/061932.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart