MAT333117121 Matrix Scientific

5-Methylthiophene-3-carbonyl chloride

  • MDL number MFCD12197866
  • Cas Number : 754190-97-1
  • Molecular Weight : 160.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061941.html

Safety Information

MSDS http://www.matrixscientific.com/061941.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart