MAT333117136 Matrix Scientific

4-Bromothiophene-2-carbonyl chloride

  • MDL number MFCD04115391
  • Cas Number : 58777-65-4
  • Molecular Weight : 225.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061956.html

Safety Information

MSDS http://www.matrixscientific.com/061956.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart