MAT333117148 Matrix Scientific

2-(2,4-Dimethylphenoxy)propanoyl chloride

  • MDL number MFCD11939548
  • Cas Number : 1114594-99-8
  • Molecular Weight : 212.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/061968.html

Safety Information

MSDS http://www.matrixscientific.com/061968.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart