MAT333117176 Matrix Scientific

(2E)-3-(3,4-Dimethoxyphenyl)acryloyl chloride

  • MDL number MFCD03840777
  • Cas Number : 39856-08-1
  • Molecular Weight : 226.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/061996.html

Safety Information

MSDS http://www.matrixscientific.com/061996.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart