MAT333117186 Matrix Scientific

4-Isopropoxy-3-methoxybenzoyl chloride

  • MDL number MFCD11940222
  • Cas Number : 3535-39-5
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 152-154�Torr
Product Link http://www.matrixscientific.com/062006.html

Safety Information

MSDS http://www.matrixscientific.com/062006.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart