MAT333117356 Matrix Scientific

2-(2-Hydroxyphenyl)-8-methylquinoline-4-carboxylic acid

  • MDL number MFCD03421945
  • Cas Number : 522596-41-4
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062176.html

Safety Information

MSDS http://www.matrixscientific.com/062176.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart