MAT333117372 Matrix Scientific

2-Ethyl-1,2,3,4-tetrahydroacridine-9-carboxylic acid

  • MDL number MFCD01924172
  • Cas Number : 351001-24-6
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062192.html

Safety Information

MSDS http://www.matrixscientific.com/062192.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart