MAT333117506 Matrix Scientific

(2E)-3-(4-Chlorophenyl)acryloyl chloride

  • MDL number MFCD00039313
  • Cas Number : 95602-71-4
  • Molecular Weight : 201.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-81
Country of Origin USA
Product Link http://www.matrixscientific.com/062326.html

Safety Information

MSDS http://www.matrixscientific.com/062326.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart