MAT333117509 Matrix Scientific

(2E)-3-(4-Methylphenyl)acryloyl chloride

  • MDL number MFCD09752196
  • Cas Number : 13565-07-6
  • Molecular Weight : 180.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/062329.html

Safety Information

MSDS http://www.matrixscientific.com/062329.html
pictograms Irritant/Stored under Argon
Availability details for
Quantity
  • Availability
  • Add to Cart