MAT333117510 Matrix Scientific

(2E)-3-(4-Methoxyphenyl)acryloyl chloride

  • MDL number MFCD00039314
  • Cas Number : 42996-84-9
  • Molecular Weight : 196.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/062330.html

Safety Information

MSDS http://www.matrixscientific.com/062330.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart