MAT333117531 Matrix Scientific

Ethyl (4-methylpiperazin-1-yl)acetate

  • MDL number MFCD01755876
  • Cas Number : 28920-67-4
  • Molecular Weight : 186.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/062351.html

Safety Information

MSDS http://www.matrixscientific.com/062351.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart