MAT333117587 Matrix Scientific

[1-Methyl-2-(5-methyl-1H-pyrazol-3-yl)ethyl]amine

  • MDL number MFCD05022480
  • Cas Number : 1025087-55-1
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062407.html

Safety Information

MSDS http://www.matrixscientific.com/062407.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart