MAT333117626 Matrix Scientific

2-Bromo-N-phenylacetamide

  • MDL number MFCD00462518
  • Cas Number : 5326-87-4
  • Molecular Weight : 214.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132
Country of Origin USA
Product Link http://www.matrixscientific.com/062446.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/062446.html
Availability details for
Quantity
  • Availability
  • Add to Cart