MAT333117717 Matrix Scientific

3-(3,4-Dichlorophenyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD03030206
  • Cas Number : 276684-04-9
  • Molecular Weight : 257.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062537.html

Safety Information

MSDS http://www.matrixscientific.com/062537.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart