MAT333117766 Matrix Scientific

trans-2-Pyrrolidin-1-ylcyclohexanol

  • MDL number MFCD19103253
  • Cas Number : 14909-81-0
  • Molecular Weight : 169.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062586.html

Safety Information

MSDS http://www.matrixscientific.com/062586.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart