MAT333117822 Matrix Scientific

(2-Oxo-2,3-dihydro-1H-indol-3-yl)acetic acid

  • MDL number MFCD09035909
  • Cas Number : 2971-31-5
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/062642.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/062642.html
Availability details for
Quantity
  • Availability
  • Add to Cart