MAT333118024 Matrix Scientific

Ethyl 4-acetyl-2-amino-5-methyl-3-furoate

  • MDL number MFCD04066410
  • Cas Number : 99076-38-7
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/062844.html

Safety Information

MSDS http://www.matrixscientific.com/062844.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart