MAT333118050 Matrix Scientific

4-(Benzylamino)phenol

  • MDL number MFCD00051312
  • Cas Number : 103-14-0
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062870.html

Safety Information

MSDS http://www.matrixscientific.com/062870.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart