MAT333118072 Matrix Scientific

Ethyl (4-oxo-3,4-dihydrophthalazin-1-yl)acetate

  • MDL number MFCD00024142
  • Cas Number : 25947-13-1
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/062892.html

Safety Information

MSDS http://www.matrixscientific.com/062892.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart