MAT333118085 Matrix Scientific

5-(4-Chlorophenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00174362
  • Cas Number : 26028-65-9
  • Molecular Weight : 211.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/062905.html

Safety Information

MSDS http://www.matrixscientific.com/062905.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart