MAT333118128 Matrix Scientific

2-{[(1,1-Dioxido-1,2-benzisothiazol-3-yl)-(1-naphthyl)amino]methyl}phenol

  • MDL number MFCD03719926
  • Cas Number : 442571-99-5
  • Molecular Weight : 414.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062948.html

Safety Information

MSDS http://www.matrixscientific.com/062948.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart