MAT333118130 Matrix Scientific

4-{[(1,1-Dioxido-1,2-benzisothiazol-3-yl)(4-methylphenyl)amino]methyl}-2-methoxyphenol

  • MDL number MFCD03270598
  • Cas Number : 433941-92-5
  • Molecular Weight : 408.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062950.html

Safety Information

MSDS http://www.matrixscientific.com/062950.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart