MAT333118134 Matrix Scientific

4-{[(4-Chlorophenyl)(1,1-dioxido-1,2-benzisothiazol-3-yl)amino]methyl}-2-methoxyphenol

  • MDL number MFCD04084649
  • Cas Number : 591242-71-6
  • Molecular Weight : 428.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062954.html

Safety Information

MSDS http://www.matrixscientific.com/062954.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart