MAT333118153 Matrix Scientific

N-(3-Hydroxyphenyl)-2-(4-oxoquinazolin-3(4H)-yl)-acetamide

  • MDL number MFCD03714289
  • Cas Number : 510760-29-9
  • Molecular Weight : 295.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/062974.html

Safety Information

MSDS http://www.matrixscientific.com/062974.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart