MAT333118156 Matrix Scientific

Methyl 4-[(methylamino)sulfonyl]benzoate

  • MDL number MFCD09754417
  • Cas Number : 474398-77-1
  • Molecular Weight : 229.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062977.html

Safety Information

MSDS http://www.matrixscientific.com/062977.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart