MAT333118163 Matrix Scientific

(2-Piperidin-3-ylethyl)amine

  • MDL number MFCD13183914
  • Cas Number : 90000-30-9
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062984.html

Safety Information

MSDS http://www.matrixscientific.com/062984.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart