MAT333118174 Matrix Scientific

5-Amino-2-cyclohexyl-1,2-dihydro-3H-pyrazol-3-one

  • MDL number MFCD13248722
  • Cas Number : 1199215-98-9
  • Molecular Weight : 181.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/062995.html

Safety Information

MSDS http://www.matrixscientific.com/062995.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart