MAT333118195 Matrix Scientific

3-Iodo-1H-1,2,4-triazole

  • MDL number MFCD00746349
  • Cas Number : 7411-20-3
  • Molecular Weight : 194.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063016.html

Safety Information

MSDS http://www.matrixscientific.com/063016.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart