MAT333118221 Matrix Scientific

6-Bromo-5-methyl-1H-indole-2,3-dione

  • MDL number MFCD00560760
  • Cas Number : 306325-13-3
  • Molecular Weight : 240.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063042.html

Safety Information

MSDS http://www.matrixscientific.com/063042.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart