MAT333118241 Matrix Scientific

N-[3-(5-Amino-1,3,4-thiadiazol-2-yl)propyl]-benzamide

  • MDL number MFCD13248729
  • Cas Number : 1199215-74-1
  • Molecular Weight : 262.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063062.html

Safety Information

MSDS http://www.matrixscientific.com/063062.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart