MAT333118245 Matrix Scientific

N-[2-(5-Amino-1,3,4-thiadiazol-2-yl)ethyl]-benzamide

  • MDL number MFCD13248733
  • Cas Number : 1199215-56-9
  • Molecular Weight : 248.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063066.html

Safety Information

MSDS http://www.matrixscientific.com/063066.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart