MAT333118258 Matrix Scientific

N-[(5-Amino-1,3,4-thiadiazol-2-yl)methyl]benzamide

  • MDL number MFCD13248746
  • Cas Number : 81837-98-1
  • Molecular Weight : 234.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215
Country of Origin USA
Product Link http://www.matrixscientific.com/063079.html

Safety Information

MSDS http://www.matrixscientific.com/063079.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart