MAT333118293 Matrix Scientific

3-(Benzoylamino)benzoic acid

  • MDL number MFCD00157843
  • Cas Number : 587-54-2
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-260
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/063114.html

Safety Information

MSDS http://www.matrixscientific.com/063114.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart