MAT333118305 Matrix Scientific

6-Methyl-1,2,4-triazine-3,5(2H,4H)-dione

  • MDL number MFCD00006457
  • Cas Number : 932-53-6
  • Molecular Weight : 127.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211-215
Country of Origin USA
Product Link http://www.matrixscientific.com/063126.html

Safety Information

MSDS http://www.matrixscientific.com/063126.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart