MAT333118480 Matrix Scientific

(3,7-Dimethyl-1H-indol-2-yl)methanol

  • MDL number MFCD15146414
  • Cas Number : 706788-99-0
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063301.html

Safety Information

MSDS http://www.matrixscientific.com/063301.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart