MAT333118633 Matrix Scientific

5-Phenyl-2H-tetrazole

  • MDL number MFCD01451271
  • Cas Number : 18039-42-4
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/063454.html

Safety Information

MSDS http://www.matrixscientific.com/063454.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart