MAT333118640 Matrix Scientific

3-Isopropyl-1H-1,2,4-triazole-5-thiol

  • MDL number MFCD15146450
  • Cas Number : 59689-64-4
  • Molecular Weight : 143.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063461.html

Safety Information

MSDS http://www.matrixscientific.com/063461.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart