MAT333118684 Matrix Scientific

5-Pyrazin-2-yl-1,3,4-oxadiazol-2-ol

  • MDL number MFCD05723092
  • Cas Number : 91144-81-9
  • Molecular Weight : 164.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063505.html

Safety Information

MSDS http://www.matrixscientific.com/063505.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart