MAT333118787 Matrix Scientific

3-Hydroxyadamantane-1-carbohydrazide

  • MDL number MFCD02208148
  • Cas Number : 68435-09-6
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173
Country of Origin USA
Product Link http://www.matrixscientific.com/063608.html

Safety Information

MSDS http://www.matrixscientific.com/063608.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart