MAT333118802 Matrix Scientific

2-(1-Adamantyl)-4-methylphenol

  • MDL number MFCD00168147
  • Cas Number : 41031-50-9
  • Molecular Weight : 242.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063623.html

Safety Information

MSDS http://www.matrixscientific.com/063623.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart