MAT333118803 Matrix Scientific

6H-Indolo[2,3-b]quinoxaline

  • MDL number MFCD00068754
  • Cas Number : 84055-81-2
  • Molecular Weight : 219.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/063624.html

Safety Information

MSDS http://www.matrixscientific.com/063624.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart