MAT333118828 Matrix Scientific

3,3'-(1,4-Phenylene)diadamantane-1-carboxylic acid

  • MDL number MFCD04128983
  • Cas Number : 113392-33-9
  • Molecular Weight : 434.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 320-323
Country of Origin USA
Product Link http://www.matrixscientific.com/063649.html

Safety Information

MSDS http://www.matrixscientific.com/063649.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart