MAT333118840 Matrix Scientific

1,4-Dimethyladamantane

  • MDL number MFCD09743483
  • Cas Number : 16267-35-9
  • Molecular Weight : 164.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/063661.html

Safety Information

MSDS http://www.matrixscientific.com/063661.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart